Learn More
Grainger Reflective Numbers And Letters 0 PK25
Catalog No. 502751221
Encompass_Preferred
The Brady Number Label is a 1 inch character height label that can be used for identification and inventory control. It features a white polyester material and permanent acrylic adhesive for application on a variety of surfaces. The labels are smudge resistant and suitable for indoor and outdoor use. This product comes in a pack of 250 labels and is offered in 6 different number sequences to choose from. The labels measure 1 inch tall by 2.75 inches long.
Catalog No. 50-275-1221 Supplier Grainger Supplier No. 14V842
The Brady Number Label is a 1 inch character height label that can be used for identification and inventory control. It features a white polyester material and permanent acrylic adhesive for application on a variety of surfaces. The labels are smudge resistant and suitable for indoor and outdoor use. This product comes in a pack of 250 labels and is offered in 6 different number sequences to choose from. The labels measure 1 inch tall by 2.75 inches long.