Learn More
Grainger Reflective Numbers And Letters E PK25
Catalog No. 502751583
Encompass_Preferred
The PK-25-1VE19 is a 1/4 HP, 115V, single phase, open drip proof motor made by Century Electric Motors. It has a totally enclosed fan cooled design with a 48 frame size, operates at 1725 RPM, and is rated for continuous duty. The motor has a 1/2" diameter shaft with a 1/4" keyway and comes with a 6 inch lead wire. It can be used for various light industrial applications.
Catalog No. 50-275-1583 Supplier Grainger Supplier No. 14V781
The PK-25-1VE19 is a 1/4 HP, 115V, single phase, open drip proof motor made by Century Electric Motors. It has a totally enclosed fan cooled design with a 48 frame size, operates at 1725 RPM, and is rated for continuous duty. The motor has a 1/2" diameter shaft with a 1/4" keyway and comes with a 6 inch lead wire. It can be used for various light industrial applications.