Learn More
Grainger Reflective Numbers And Letters P PK25
Catalog No. 502751098
Encompass_Preferred
The Brady Letter Label 1 in Character Height, White on Black Vinyl Label features 1-inch tall black characters on a white background. Ideal for identifying wires, panels, components, equipment, tools, and materials, it comes in a roll format for easy dispensing and application. It has a strong permanent adhesive that sticks to various surfaces. The durable vinyl is abrasion and chemical resistant, and withstands extreme temperatures. With high contrast print, it ensures excellent readability. The Brady Letter Label offers an effective and economical solution for clear identification in industrial settings.
Catalog No. 50-275-1098 Supplier Grainger Supplier No. 14V831
Add to Cart
Add to Cart
The Brady Letter Label 1 in Character Height, White on Black Vinyl Label features 1-inch tall black characters on a white background. Ideal for identifying wires, panels, components, equipment, tools, and materials, it comes in a roll format for easy dispensing and application. It has a strong permanent adhesive that sticks to various surfaces. The durable vinyl is abrasion and chemical resistant, and withstands extreme temperatures. With high contrast print, it ensures excellent readability. The Brady Letter Label offers an effective and economical solution for clear identification in industrial settings.