Learn More
Grainger Reflective Numbers And Letters L PK25
Catalog No. 502751104
Encompass_Preferred
The Stranco Inc. Reflective Letter Label 2-8CL50 is a reflective letter label made of vinyl with an adhesive backing. It has 2 inch tall letters and numbers in black on a silver reflective background. The label comes in a roll that is 50 feet long and can be cut to desired lengths. It is designed for indoor or outdoor use on smooth flat surfaces and has a service temperature range of -40°F to 200°F. This reflective label provides high visibility and is useful for identifying equipment, tools, storage areas, aisles, and more in warehouses, factories, construction sites and other commercial and industrial locations.
Catalog No. 50-275-1104 Supplier Grainger Supplier No. 14V827
The Stranco Inc. Reflective Letter Label 2-8CL50 is a reflective letter label made of vinyl with an adhesive backing. It has 2 inch tall letters and numbers in black on a silver reflective background. The label comes in a roll that is 50 feet long and can be cut to desired lengths. It is designed for indoor or outdoor use on smooth flat surfaces and has a service temperature range of -40°F to 200°F. This reflective label provides high visibility and is useful for identifying equipment, tools, storage areas, aisles, and more in warehouses, factories, construction sites and other commercial and industrial locations.