Learn More
Grainger Reflective Numbers And Letters 0 PK25
Catalog No. 502752407
Encompass_Preferred
The Brady Number Label is a 1 inch character height label made of vinyl material. It has a white background with black print and is suitable for indoor use. The label comes in sheets of 48 labels and can be used with the Brady TLS 2200 Thermal Transfer Printer. It is designed for wire and cable marking, panel and equipment ID, and general purpose identification. The vinyl material is durable and abrasion resistant. With high contrast print, it offers excellent readability that complies with ANSI and OSHA standards. The 1 inch tall characters allow visibility from distances. This label provides an organized and professional wire and cable marking solution.
Catalog No. 50-275-2407 Supplier Grainger Supplier No. 14V804
The Brady Number Label is a 1 inch character height label made of vinyl material. It has a white background with black print and is suitable for indoor use. The label comes in sheets of 48 labels and can be used with the Brady TLS 2200 Thermal Transfer Printer. It is designed for wire and cable marking, panel and equipment ID, and general purpose identification. The vinyl material is durable and abrasion resistant. With high contrast print, it offers excellent readability that complies with ANSI and OSHA standards. The 1 inch tall characters allow visibility from distances. This label provides an organized and professional wire and cable marking solution.