Learn More
Brady™ Reflective sheeting laminated to Aluminum Sign: 45 MIN VEHICLE CHARGING ONLY, 7 AM-10 PM MON-FRI
Catalog No. 19111542 Shop All Brady Corporation Products
Change view
Click to view available options
Includes:
Dual Arrow
Left Arrow
Right Arrow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-111-542 Left Arrow
19-111-541 Dual Arrow
19-111-540 Right Arrow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-111-542 Supplier Brady™ Supplier No. 141690
  • $102.00
    $72.65 / Each of 1
    Save $29.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign with reflective sheeting laminate is a durable, clear and effective tool to designate electric vehicle parking and charging locations.
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Includes Left Arrow
Material Aluminum
Shape Rectangular
Dimensions (L x W) 12 x 18 in.
Displayed Text 45 MIN VEHICLE CHARGING ONLY, 7 AM-10 PM MON-FRI
Color Green on White
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.