Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Reflective Sheeting Sign Kit: Bradylite NFPA Diamonds

Catalog No. 19110395 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
5 x 5 in.
10 x 10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-110-395 10 x 10 in.
19-110-394 5 x 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-110-395 Supplier Brady™ Supplier No. 58274
  • $350.50
    $252.65 / Pack of 10

    Save $97.85

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Improve workplace safety with proper signage and markings. This Brady™ reflective sheeting sign kit is a durable, clear and effective tool to raise awareness to workplace hazards.

These Bradylite™ reflective numbers and letters are designed for use in low-light or dark areas, offering reflection 100 times brighter than a plain white surface. They are constructed from Engineering Grade B-997 reflective sheeting and feature an aggressive, permanent acrylic adhesive suitable for cold temperature application.

  • Material: Engineering Grade B-997 Reflective Sheeting
  • Dimensions: 2 7/8 in. H x 1 3/4 in. W (2 1/2 in. character height)
  • Service Temperature Range: -40°F to 200°F
  • Minimum Application Temperature: -17°F
  • Resistance Properties: Abrasion-resistant and chemical-resistant
  • Durability: 4 to 6 years average outdoor durability
  • Applications: Facility, hazard, and safety identification
  • Compliance: REACH SVHC compliant; RoHS not applicable

Specifications

Material Reflective Sheeting
Mounting Type Self-adhesive
Shape Square
Dimensions (L x W) 10 x 10 in.
Displayed Text Reflective Sheeting Sign Kit: Bradylite NFPA Diamonds
Product Type NFPA Placard
Color Yellow
WARNING: Cancer - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.