Learn More
Grainger Reflective Symbol Dash 3-1/2 in H PK10
Catalog No. 502753118
Encompass_Preferred
The Brady Symbol Label is a 3 inch character height label that features a high performance acrylic adhesive for bonding to low surface energy plastics such as polypropylene and powder coated paints. It has a temperature range of -40°F to 302°F and is made from vinyl. This label comes in white and is ideal for barcoding, product identification, and general purpose labeling applications. It meets the BS5609 certification for marine environments.
Catalog No. 50-275-3118 Supplier Grainger Supplier No. 14R052
The Brady Symbol Label is a 3 inch character height label that features a high performance acrylic adhesive for bonding to low surface energy plastics such as polypropylene and powder coated paints. It has a temperature range of -40°F to 302°F and is made from vinyl. This label comes in white and is ideal for barcoding, product identification, and general purpose labeling applications. It meets the BS5609 certification for marine environments.