Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Reflectoquant™ Ammonium Test Strips

Catalog No. M1168990001 Shop All MilliporeSigma Products
Change view
Click to view available options
Graduations:
0.2 to 7mg/L
5 to 20mg/L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Graduations
M1168990001 5 to 20mg/L
M1168920001 0.2 to 7mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1168990001 Supplier MilliporeSigma™ Supplier No. 1.16899.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with the Reflectoquant™ system

For accurate and precise on-site analysis.

  • Available using the indophenol blue or the Nessler method
  • Use in agriculture, food or water analysis, hygiene monitoring, industrial analysis

Order Info

50 tests per kit

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) The test measures both ammonium ions and dissolved ammonia in e.g. groundwater and surface water, seawater, drinking water and aquarium water
Graduations 5 to 20mg/L
Quantity 50 Tests
Storage Requirements 2°C to 8°C
Test Method Indophenol blue
Type Ammonium Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.