Learn More
Roboz Surgical Roboz Surgical Instruments, Wound Clips, 7mm, Stainless Steel, 1000/Pk.
SDP
Catalog No. NC9884167
Encompass
These stainless steel 7mm wound clips are engineered for routine wound closure in laboratory animal research settings. The clips provide a secure and reliable closure for surgical incisions.
  • Manufactured from stainless steel
  • Compatible with mri procedures
  • Non-toxic and non-pyrogenic material composition
  • Designed for exclusive use with 7mm wound clip appliers
  • Supplied in a convenient box of 1000 units
  • Intended solely for animal research applications
Catalog No. NC9884167 Supplier Roboz Surgical Supplier No. RS9255
These stainless steel 7mm wound clips are engineered for routine wound closure in laboratory animal research settings. The clips provide a secure and reliable closure for surgical incisions.
  • Manufactured from stainless steel
  • Compatible with mri procedures
  • Non-toxic and non-pyrogenic material composition
  • Designed for exclusive use with 7mm wound clip appliers
  • Supplied in a convenient box of 1000 units
  • Intended solely for animal research applications