Learn More
Stoelting™ Reflex Wound Clip Remover
SDP
Catalog No. 10001027 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Removing Clips
Catalog No. For Use With (Application)
10-001-027 Removing Clips
1 options
Catalog No. 10-001-027 Supplier Stoelting™ Supplier No. 59037
Add to Cart
Add to Cart
Wound clips allow for smaller, rapid suturing.
  • 7mm clips are an ideal size for mouse surgery
  • Clips are sold in individual packs of 20, unsterilized
  • Applier can easily handle one full pack of clips
  • Autoclavable
  • Instruments are for non-clinical use only (not for human use)
Specifications
For Use With (Application) Removing Clips
For Use With (Equipment) 7 mm clips