Learn More
Stoelting™ Reflex Wound Clips
SDP
Catalog No. 10001024 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
7 mm
Catalog No. Size
10-001-024 7 mm
1 options
Catalog No. 10-001-024 Supplier Stoelting™ Supplier No. 59034
Add to Cart
Add to Cart
Wound clips allow for smaller, rapid suturing.
  • 7mm clips are an ideal size for mouse surgery
  • Clips are sold in individual packs of 20, unsterilized
  • Applier can easily handle one full pack of clips
  • Autoclavable
  • Instruments are for non-clinical use only (not for human use)
Specifications
Size 7 mm
Quantity 1000 wound clips