Learn More
Stoelting™ Reflex Wound Closures
SDP
Catalog No. 10001028 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
7 mm
Catalog No. Size
10-001-028 7 mm
1 options
Catalog No. 10-001-028 Supplier Stoelting™ Supplier No. 59038
Add to Cart
Add to Cart
Stoelting™ Reflex Wound Closure Clips allow for smaller, rapid suturing.
  • Ideal size for mouse surgery
  • Sold in individual packs of 20
  • Unsterilized
  • User can easily handle one full pack of clips
  • Autoclavable
Specifications
For Use With (Application) Neuroscience
Size 7 mm
Animal Mouse