Learn More
MOLDPRO INC Reflux cap for 50 mL Digestion Tube, clear, 1000/pkg, PP Homopolymer
Catalog No. NC1328476
Encompass_Preferred
MoldPro MP-112 Reflux Cap is designed to fit all 50 mL digestion vessels. Features center hole for convenient access during digestions. • Made from virgin polypropylene • 1000 pieces/case • Direct replacement for Environmental Express SC506
Catalog No. NC1328476 Supplier MOLDPRO INC Supplier No. MP112
MoldPro MP-112 Reflux Cap is designed to fit all 50 mL digestion vessels. Features center hole for convenient access during digestions. • Made from virgin polypropylene • 1000 pieces/case • Direct replacement for Environmental Express SC506