Learn More
Chemglass Life Sciences Condenser, Reflux, 205mm, 14/20 Joint, Removable Hose Connections, 100mm Jacket Length
SDP
Catalog No. CG1213HC01
Encompass_Preferred
Condenser, Reflux, 205mm, 14/20 Joint, Removable Hose Connections, 100mm Jacket Length. Reflux condenser similar to CG-1213 but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Hose connections have an O.D. of 8mm and an I.D. of 4mm. Supplied complete with two GL-14 open top screw caps. See CG-198 for replacement GL fittings.
Catalog No. CG1213HC01 Supplier Chemglass Life Sciences Supplier No. CG1213HC01
Condenser, Reflux, 205mm, 14/20 Joint, Removable Hose Connections, 100mm Jacket Length. Reflux condenser similar to CG-1213 but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Hose connections have an O.D. of 8mm and an I.D. of 4mm. Supplied complete with two GL-14 open top screw caps. See CG-198 for replacement GL fittings.