Learn More
Chemglass Life Sciences Condenser, Reflux, 315mm, 24/40 Joint, Removable Hose Connections, 150mm Jacket Length
SDP
Catalog No. CG1213HC09
Encompass_Preferred
Condenser, Liebig, 325mm, 29/42 Joint, 200mm Jacket Length. Condenser has a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG1213HC09 Supplier Chemglass Life Sciences Supplier No. CG1213HC09
Condenser, Liebig, 325mm, 29/42 Joint, 200mm Jacket Length. Condenser has a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.