Learn More
Chemglass Life Sciences Condenser, Reflux, 415mm, 24/40 Joint, Style B, 250mm Jacket Length
SDP
Supplier:  Chemglass Life Sciences CG121303
Encompass_Preferred
Condenser, Reflux, 415mm, 24/40 Joint, Style B, 250mm Jacket Length. Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG121303