Learn More
Chemglass Life Sciences Condenser, Reflux, 465mm, 24/40 Joint, Style B, 300mm Jacket Length
SDP
Catalog No. CG121305
Encompass_Preferred
Condenser, Reflux, 465mm, 24/40 Joint, Style B, 300mm Jacket Length. Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG121305 Supplier Chemglass Life Sciences Supplier No. CG121305
Condenser, Reflux, 465mm, 24/40 Joint, Style B, 300mm Jacket Length. Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.