Learn More
Chemglass Life Sciences Condenser, High Efficiency, 730mm, 45/50 Joint, 600mm Jacket Length, 100mm OD Jacket
SDP
Catalog No. CG1215C02
Encompass_Preferred
Condenser, High Efficiency, 730mm, 45/50 Joint, 600mm Jacket Length. This unique design combines some of the best features for refluxing in a single condenser. Coolant, introduced into the upper hose connection, fills a large cold finger that is wrapped with coils. After the coolant travels through the coils, it enters the water jacket to maximize cooling surface along the vapor path. Angled take off joint is a standard taper 24/40, and the angled take off joint for the 45/50 size is a standard taper 29/42. Hose connections for the 24/40 size condenser are 10mm at the largest serration. Hose connections for the 45/50 size are 15mm at the largest serration.
Catalog No. CG1215C02 Supplier Chemglass Life Sciences Supplier No. CG1215C02
Condenser, High Efficiency, 730mm, 45/50 Joint, 600mm Jacket Length. This unique design combines some of the best features for refluxing in a single condenser. Coolant, introduced into the upper hose connection, fills a large cold finger that is wrapped with coils. After the coolant travels through the coils, it enters the water jacket to maximize cooling surface along the vapor path. Angled take off joint is a standard taper 24/40, and the angled take off joint for the 45/50 size is a standard taper 29/42. Hose connections for the 24/40 size condenser are 10mm at the largest serration. Hose connections for the 45/50 size are 15mm at the largest serration.