Learn More
Plastilite Corp Mailing and Shipping Containers, Insulated Containers. EPS Shipper. Foam Insulated Container. Foam Shipper. Insulated Shipper. Insulated Shipper Container. Biodegradable REfoam insulated shipper. RSL816-C36 cooler in box.
Catalog No. NC3584382
Encompass_Preferred
Mailing and Shipping Containers, Insulated Containers. EPS Shipper. Foam Insulated Container. Foam Shipper. Insulated Shipper. Insulated Shipper Container. Temperature-controlled Package Shipper. Biodegradable REfoam insulated shipper molded with up to 30% recycled EPS content. 1.5” thick-wall REfoam cooler with white outer box at 32ECT strength with “PERISHABLE” and up arrows on two sides. Durable and highly insulating for your temperature sensitive products such as pharmaceutical and biological materials. Use with dry ice, gel ice packs, foam brick ice packs, and PCMs as refrigerant material. With proper amounts of refrigerant material this cooler can protect up to four days depending on ambient shipping temperatures. RSL816-C36 cooler in box, 1.5” thick. 36/PK. Inside Dimensions of 12.125"x10.625"x8.625" offering .64 cubic foot payload capacity. This EPS REfoam cooler is recyclable at more than 400 locations across NA.
Catalog No. NC3584382 Supplier Plastilite Corp Supplier No. RSL816C36
Mailing and Shipping Containers, Insulated Containers. EPS Shipper. Foam Insulated Container. Foam Shipper. Insulated Shipper. Insulated Shipper Container. Temperature-controlled Package Shipper. Biodegradable REfoam insulated shipper molded with up to 30% recycled EPS content. 1.5” thick-wall REfoam cooler with white outer box at 32ECT strength with “PERISHABLE” and up arrows on two sides. Durable and highly insulating for your temperature sensitive products such as pharmaceutical and biological materials. Use with dry ice, gel ice packs, foam brick ice packs, and PCMs as refrigerant material. With proper amounts of refrigerant material this cooler can protect up to four days depending on ambient shipping temperatures. RSL816-C36 cooler in box, 1.5” thick. 36/PK. Inside Dimensions of 12.125"x10.625"x8.625" offering .64 cubic foot payload capacity. This EPS REfoam cooler is recyclable at more than 400 locations across NA.