Learn More
Anatech Ltd REFORM FORMALIN NBF 6/CS
Catalog No. NC9175207
Encompass_Preferred
Re-Form NBF; Phosphate salt solution to make neutral buffered formalin; Each quart is sufficient for approximately 10 gal.; 6/qt.
Catalog No. NC9175207 Supplier Anatech Ltd Supplier No. 235
Re-Form NBF; Phosphate salt solution to make neutral buffered formalin; Each quart is sufficient for approximately 10 gal.; 6/qt.