Learn More
Julabo USA Inc CP-200F Refrigerated/heating circulator -20...+200 °C; 200W cooling @ 20 °C; 1 kW heater; 3-4 L bath tank; adjustable flow pump 8-27L/min; 1.5-10.2 psi with 8 and 12 mm hose barbs; USB and RS232 ports
SDP
Supplier:  Julabo USA Inc 9013701.02
Encompass_Preferred
CORIO™ CP, The new generation of laboratory circulators from JULABO.CORIO ™ CP refrigerated/heating circulators are suitable for temperature applications with circulation to an external apparatus or immersing a device in the bath tank. The CORIO™ CP circulator incorporates easy change-over for control of fluid circulation internally or externally. Unit fittings M16x1 male threaded adapters including a pair of 8 and 12 mm hose barbs. A range of models provide temperatures spanning -50...+200 °C with cooling capacity up to 1 kW at 20 °C.Unit fittings M16x1 male threaded adapters including a pair of 8 and 12 mm hose barbs. Systems include insulated stainless-steel tanks, lid and integrated drain port.Bright LED display with easy to use interface. USB and RS232 connections for external PC control and data-logging; PID temperature control provides ±0.03 °C stability. Adjustable centrifugal pump capacity of 8-27 L/min with pressure from 1.5-10.2 psi.
Catalog No. 50-190-4657
Only null left
Add to Cart
Request a Quote
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov