Learn More
Anton Paar REFRACTOMETER REFERENCE LIQUID 20ML WATER, 1.3329ND, WORKS CERTIFICATE
Catalog No. NC2736777
Encompass_Preferred
Compatible with: Abbemat 200 Economy Line Refractometer, Abbemat 3000, Abbemat 3100, Abbemat 3200, Brix refractometer: Abbemat Juice Station, Abbemat Performance 300, Abbemat Performance 500, Abbemat Performance Plus 350, Abbemat Performance Plus 550, Abbemat 450, Abbemat 650, Multiwavelengths Refractometer: Abbemat MW
Catalog No. NC2736777 Supplier Anton Paar Supplier No. 46928
Compatible with: Abbemat 200 Economy Line Refractometer, Abbemat 3000, Abbemat 3100, Abbemat 3200, Brix refractometer: Abbemat Juice Station, Abbemat Performance 300, Abbemat Performance 500, Abbemat Performance Plus 350, Abbemat Performance Plus 550, Abbemat 450, Abbemat 650, Multiwavelengths Refractometer: Abbemat MW