Learn More
Waters Corp Waters Refractive Index Performance and Operation Solution, Liquid, 10 mL, System Performance
Catalog No. 50818870 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Refractive Index Performance and Operation Solution is a standard used for the operation and performance assessment of Refractive Index (RI) detectors. This solution is also referred to as a “universal detector” in liquid chromatography because of its ability to detect all chemicals. It is shipped in liquid form and requires special handling due to hazardous material content. It has a shelf life of 36 months and should be stored at room temperature.
  • Standard for operation and performance assessment of Refractive Index detectors
  • Functions as a universal detector in liquid chromatography
  • Manufactured in facilities adhering to cGMP and ISO 9001 guidelines
  • Manufacturing process is meticulously monitored and controlled
  • Contains 2.0 g/L caffeine ±1% standard solution
  • Requires special freight handling as it contains hazardous material
Catalog No. 50-818-870 Supplier Waters Corp Supplier No. 700001713
The Refractive Index Performance and Operation Solution is a standard used for the operation and performance assessment of Refractive Index (RI) detectors. This solution is also referred to as a “universal detector” in liquid chromatography because of its ability to detect all chemicals. It is shipped in liquid form and requires special handling due to hazardous material content. It has a shelf life of 36 months and should be stored at room temperature.
  • Standard for operation and performance assessment of Refractive Index detectors
  • Functions as a universal detector in liquid chromatography
  • Manufactured in facilities adhering to cGMP and ISO 9001 guidelines
  • Manufacturing process is meticulously monitored and controlled
  • Contains 2.0 g/L caffeine ±1% standard solution
  • Requires special freight handling as it contains hazardous material