Learn More
PCE AMERICAS INC Porable Pocket Brix Refractometer 0 to 32% Brix (Sucrose or Sugar Content)
SDP
Catalog No. NC3654409
Encompass_Preferred
"PCE-018 is a handheld refractometer used to determine the % Brix (sucrose or sugar content) of low-concentration translucent liquids such as fruit and vegetable juices, maple sap and industrial coolants. This portable pocket Brix refractometer is specially designed to provide high-accuracy readings.The ultra-smooth focusing eyepiece provides high-quality optics, presenting an easy-to-read scale on a semi-transparent blue and fully transparent clear background. Ambient room lighting offers ample illumination to clearly read the results.
Catalog No. NC3654409 Supplier PCE AMERICAS INC Supplier No. PCE018
"PCE-018 is a handheld refractometer used to determine the % Brix (sucrose or sugar content) of low-concentration translucent liquids such as fruit and vegetable juices, maple sap and industrial coolants. This portable pocket Brix refractometer is specially designed to provide high-accuracy readings.The ultra-smooth focusing eyepiece provides high-quality optics, presenting an easy-to-read scale on a semi-transparent blue and fully transparent clear background. Ambient room lighting offers ample illumination to clearly read the results.