Learn More
Electron Microscopy Sciences Waterproof Refractometer Brix
SDP
Supplier:  Electron Microscopy Sciences 72080
Encompass_Preferred
Cover the entire 0 to 95 percent Brix range with one instrument, along with refractive index, temperature, and dextran, fructose, glucose, lactose, and maltose scales. Housed in a protective rubberized IP65 Waterproof case to safeguard the electronics from wet environments, which also makes for easy clean ups. A scratch proof sapphire lens ensures continued accuracy over time.Measure samples as small as 1 ml, in the field, with laboratory accuracy. Sample temperature is initially displayed on the large LCD followed by the measurement results. The sophisticated microprocessor provides automatic calibration, automatic temperature compensation and automatic shutoff (after 3 minutes). Features one button calibration to distilled water and indicates when the battery is low. Powered by a rechargeable, long-life, lithium ion battery providing 50,000 tests or about half a year use on a single charge. Includes instructions, carrying case, transfer-pipet and an AC power adapter.
Catalog No. 50-192-9319