Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific Refractometer Contact Solution (Bromonaphthalene), NIST

Catalog No. 14127338
Change view
Click to view available options
Refractive Index:
1.65 nD
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Refractive Index
14-127-338 1.65 nD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-127-338 Supplier Heathrow Scientific Supplier No. 45000OS001

missing translation for 'validMainframeMsg'

Refractometer Standard, Contact Solution

  • Color: Clear
  • Quantity: 1 Ea.
  • Warranty: 1 Year

Specifications

Type Portable Refractometer
Refractive Index 1.65 nD
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.