Learn More
Brookfield Engineer Labs Circulating water bath, -20°C +170°C
Catalog No. NC0346195
Encompass
Provides standalone operation - no tap water requiredEasy control of set-pointConfigured to measure viscosity directly in the bath - accommodates 600 mL beakerBuilt-in circulator pumps to external devices, Controller Cooling Temperature StabilityModel; TC-550SD, Temp Range; -20°C +170°C, Controller; SD
Catalog No. NC0346195 Supplier Brookfield Engineer Labs Supplier No. TC550SD115
Add to Cart
Add to Cart
Request a Quote
Provides standalone operation - no tap water requiredEasy control of set-pointConfigured to measure viscosity directly in the bath - accommodates 600 mL beakerBuilt-in circulator pumps to external devices, Controller Cooling Temperature StabilityModel; TC-550SD, Temp Range; -20°C +170°C, Controller; SD