Learn More
Polyscience 7L Space-Saving Refrigerated Circulator, Advanced Programmable Controller (-40 to 200°C); 120V, 60Hz
SDP
Catalog No. NC0838449
Encompass_Preferred
PolyScience™ 7L Space-Saving Refrigerated CirculatorAdvanced Programmable Controller-40 to 200°C Working Temperature RangeTemperature Stability: ±0.01°CCooling Capacity at 20°C: 360W 1100W HeaterMaximum Pressure: 4.3 psi (0.30 bar) Maximum Pressure Flow Rate:5.3 gpm (20.1 LPM) 120V, 60HzFeatures:• OpenMode time time/temperature programming (no restrictions on number of programs or steps)• Intuitive 4.3” (10.9 cm) SmartTouch display• 11 languages: French, German, Spanish, Chinese, Portuguese, Russian, Hindi, Arabic, Italian, Korean, English• Variable speed pump with open- and closed-loop external circulation capability• Swivel 180™ Rotating Controller• On-board connectivity: USB-A & B, Ethernet, RS-232/RS-485, remote on/off and external temperature probe• Event scheduling (time & date) with real-time clock
Catalog No. NC0838449 Supplier Polyscience Supplier No. AP07R40A11B
Add to Cart
Add to Cart
Request a Quote
PolyScience™ 7L Space-Saving Refrigerated CirculatorAdvanced Programmable Controller-40 to 200°C Working Temperature RangeTemperature Stability: ±0.01°CCooling Capacity at 20°C: 360W 1100W HeaterMaximum Pressure: 4.3 psi (0.30 bar) Maximum Pressure Flow Rate:5.3 gpm (20.1 LPM) 120V, 60HzFeatures:• OpenMode time time/temperature programming (no restrictions on number of programs or steps)• Intuitive 4.3” (10.9 cm) SmartTouch display• 11 languages: French, German, Spanish, Chinese, Portuguese, Russian, Hindi, Arabic, Italian, Korean, English• Variable speed pump with open- and closed-loop external circulation capability• Swivel 180™ Rotating Controller• On-board connectivity: USB-A & B, Ethernet, RS-232/RS-485, remote on/off and external temperature probe• Event scheduling (time & date) with real-time clock