Learn More
Felix Storch Inc ACCUCOLD REFRIGERATOR 33.5H x 23.63W x 23.5D
SDP
Catalog No. NC2087852
Encompass_Preferred
The FF6LW is a counter height all-refrigerator with a full 5.5 cu.ft. capacity inside a slim 24" footprint. Designed for freestanding use, this model has a white exterior and user-reversible door swing. A front-mounted lock offers added security.Inside, the FF6LW utilizes low maintenance automatic defrost operation. Our unique "hidden" evaporator is set behind the rear wall to create a seamless, easy-to-clean interior that also maximizes shelf space to provide generous storage. Adjustable wire shelves allow users to reconfigure the interior to accommodate a variety of sizes. Additional features include automatic interior lighting, door storage, and a crisper drawer. This unit includes an adjustable dial thermostat: https://www.accucold.com/product/FF6LW
This item is not returnable. View return policy
Catalog No. NC2087852 Supplier Felix Storch Inc Supplier No. FF6LW
The FF6LW is a counter height all-refrigerator with a full 5.5 cu.ft. capacity inside a slim 24" footprint. Designed for freestanding use, this model has a white exterior and user-reversible door swing. A front-mounted lock offers added security.Inside, the FF6LW utilizes low maintenance automatic defrost operation. Our unique "hidden" evaporator is set behind the rear wall to create a seamless, easy-to-clean interior that also maximizes shelf space to provide generous storage. Adjustable wire shelves allow users to reconfigure the interior to accommodate a variety of sizes. Additional features include automatic interior lighting, door storage, and a crisper drawer. This unit includes an adjustable dial thermostat: https://www.accucold.com/product/FF6LW