Learn More
AMUZA INC TOMY MDX-310 High Speed Refrigerated Centrifuge; 15000 rpm; 21, 890G
SDP
Catalog No. NC3711170
Encompass
TOMY MDX-310 High-Speed Refrigerated Centrifuge has a stackable “rack-in-rotor” system for easy, instantaneous swapping of rotors. There is no need to pull out tools or buy several centrifuges to accommodate your large array of tubes and sample media.Accommodates everything from 1.5 / 2.0 mL tubes to 96-well plates to 50 mL tubesStack up to 3 racks (72 – 1.5/2.0 mL microtubes in one cycle).High power – Runs at up to 15,000 RPM / 21,890G.Superior safety qualities – The electronic lid interlock system prevents the lid from opening while in operation. Smart sensing systems are in place for lid opening/closing, rotor imbalance, excessive speed, motor over-current (with circuit breaker), and abnormal chamber temperature.Eco-mode reduces power consumption by up to 43%, compared to earlier models.
Catalog No. NC3711170 Supplier AMUZA INC Supplier No. MDX310
Add to Cart
Add to Cart
Request a Quote
TOMY MDX-310 High-Speed Refrigerated Centrifuge has a stackable “rack-in-rotor” system for easy, instantaneous swapping of rotors. There is no need to pull out tools or buy several centrifuges to accommodate your large array of tubes and sample media.Accommodates everything from 1.5 / 2.0 mL tubes to 96-well plates to 50 mL tubesStack up to 3 racks (72 – 1.5/2.0 mL microtubes in one cycle).High power – Runs at up to 15,000 RPM / 21,890G.Superior safety qualities – The electronic lid interlock system prevents the lid from opening while in operation. Smart sensing systems are in place for lid opening/closing, rotor imbalance, excessive speed, motor over-current (with circuit breaker), and abnormal chamber temperature.Eco-mode reduces power consumption by up to 43%, compared to earlier models.