Learn More
Polyscience 7L Space-Saving Refrigerated Circulator, Advanced Digital Controller (-20 to 200°C); 120V, 60Hz
SDP
Catalog No. NC1040122
Encompass_Preferred
PolyScience™ 7 Liter Space-Saving Refrigerated CirculatorAdvanced Digital Controller-20 to 200°C Working Temperature RangeTemperature Stability: ±0.01°CCooling Capacity at 20°C: 200W 1100W HeaterMaximum Pressure: 4.3 psi (0.30 bar) Maximum Pressure Flow Rate: 5.3 gpm (20.1 LPM) 120V, 60HzFeatures:• Intuitive 3.75” (9.5 cm) display with touch-pad control• 4 languages: French, German, Spanish, English• Swivel 180™ Rotating Controller• On-board connectivity: USB-A & B, Ethernet, RS-232/RS-485, and external temperature probe• On-screen prompts• Automatic performance optimization and specific heat tuning• Single-point calibration capability
Catalog No. NC1040122 Supplier Polyscience Supplier No. AD07R20A11B
Add to Cart
Add to Cart
Request a Quote
PolyScience™ 7 Liter Space-Saving Refrigerated CirculatorAdvanced Digital Controller-20 to 200°C Working Temperature RangeTemperature Stability: ±0.01°CCooling Capacity at 20°C: 200W 1100W HeaterMaximum Pressure: 4.3 psi (0.30 bar) Maximum Pressure Flow Rate: 5.3 gpm (20.1 LPM) 120V, 60HzFeatures:• Intuitive 3.75” (9.5 cm) display with touch-pad control• 4 languages: French, German, Spanish, English• Swivel 180™ Rotating Controller• On-board connectivity: USB-A & B, Ethernet, RS-232/RS-485, and external temperature probe• On-screen prompts• Automatic performance optimization and specific heat tuning• Single-point calibration capability