Learn More
Felix Storch Inc ACCUCOLD REFRIG UPRGHT MAN DEF 18F
SDP
Catalog No. NC3780078
Encompass_Preferred
Model URM19W is an upright all-refrigerator with an 18.3 cu.ft. interior storage capacity. Manual defrost operation ensures a more stable interior with none of the warming cycles that can compromise temperature in traditional frost-free cooling systems. The dial thermostat is located inside to prevent accidental temperature adjustments, with a thermometer/alarm mounted externally to ensure a readout of the current and high/low interior temperature is visible without opening the door. The temperature probe is encased in glass beads and positioned in a fixed shelf in the center of the unit to best simulate the temperature of stored items. We include an additional probe hole to accommodate additional monitoring equipmen: https://www.accucold.com/product/URM19W
Catalog No. NC3780078 Supplier Felix Storch Inc Supplier No. URM19W
Model URM19W is an upright all-refrigerator with an 18.3 cu.ft. interior storage capacity. Manual defrost operation ensures a more stable interior with none of the warming cycles that can compromise temperature in traditional frost-free cooling systems. The dial thermostat is located inside to prevent accidental temperature adjustments, with a thermometer/alarm mounted externally to ensure a readout of the current and high/low interior temperature is visible without opening the door. The temperature probe is encased in glass beads and positioned in a fixed shelf in the center of the unit to best simulate the temperature of stored items. We include an additional probe hole to accommodate additional monitoring equipmen: https://www.accucold.com/product/URM19W