Learn More
Julabo USA Inc Refrigerated/heating circulator, JULABO, DYNEO DD-449F, 115V/60Hz, R290 Refrigerant. -32…200C. 1kW heater. Cooling power 440W at 20C, USB-B I/O, External Pt100, programmable. USB-A data-logging. 20-26L tank, Recommended for internal application.
SDP
Catalog No. 5000693582
Encompass_Preferred
Refrigerated/heating circulator, DYNEO DD-449F, 115V/60Hz, R290 Refrigerant. -32…200C. 1kW heater. Cooling power 440W at 20C 350W at 0C 270W at -10C 200W at -20C 60W at -30C. Adjustable centrifugal pump 8-27LPM, 1.5-11.6 PSI. USB-A data-logging; USB-B external communication. Lemosa Pt100 port for external temperature control and monitoring. Color screen with click and scroll navigation. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 20...26L bath tank. Bath tank opening: 11x13.8/7.9 in (WxL/D); Dimensions: 14.6 x 23.2 x 27.2 in. 87.1lbs. Recommended for internal bath applications.
Catalog No. 50-006-93582 Supplier Julabo USA Inc Supplier No. 9021716.N1.22
Refrigerated/heating circulator, DYNEO DD-449F, 115V/60Hz, R290 Refrigerant. -32…200C. 1kW heater. Cooling power 440W at 20C 350W at 0C 270W at -10C 200W at -20C 60W at -30C. Adjustable centrifugal pump 8-27LPM, 1.5-11.6 PSI. USB-A data-logging; USB-B external communication. Lemosa Pt100 port for external temperature control and monitoring. Color screen with click and scroll navigation. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 20...26L bath tank. Bath tank opening: 11x13.8/7.9 in (WxL/D); Dimensions: 14.6 x 23.2 x 27.2 in. 87.1lbs. Recommended for internal bath applications.