Learn More
Julabo USA Inc Refrigerated/heating circulator, JULABO, DYNEO DD-800F.D, 115V/60Hz, R1270 Refrigerant. -40…200C. 1kW heater. Cooling power 840W at 20C, USB-B and RS232 I/O, External Pt100, programmable. USB-A data-logging.
SDP
Catalog No. 5000693599
Encompass_Preferred
Refrigerated/heating circulator, DYNEO DD-800F.D, 115V/60Hz, R1270 Refrigerant. -40…200C. 1kW heater. Cooling power 840W at 20C 740W at 0C 390W at -20C 240W at -30C 100W at -40C. Adjustable centrifugal pump 8-27LPM, 1.5-11.6 PSI. USB-A data-logging; USB-B and RS232 external communication. Lemosa Pt100 port for external temperature control and monitoring. Color screen with click and scroll navigation. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 5...7.5L bath tank. Bath tank opening: 7.1x5.1/5.9 in (WxL/D); Dimensions: 13 x 18.5 x 27.6 in. 92.6lbs
Catalog No. 50-006-93599 Supplier Julabo USA Inc Supplier No. 9021715.N1.22.D
Refrigerated/heating circulator, DYNEO DD-800F.D, 115V/60Hz, R1270 Refrigerant. -40…200C. 1kW heater. Cooling power 840W at 20C 740W at 0C 390W at -20C 240W at -30C 100W at -40C. Adjustable centrifugal pump 8-27LPM, 1.5-11.6 PSI. USB-A data-logging; USB-B and RS232 external communication. Lemosa Pt100 port for external temperature control and monitoring. Color screen with click and scroll navigation. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 5...7.5L bath tank. Bath tank opening: 7.1x5.1/5.9 in (WxL/D); Dimensions: 13 x 18.5 x 27.6 in. 92.6lbs