Learn More
Julabo USA Inc Refrigerated/heating circulator, JULABO, MAGIO MS-1200FW, water-cooled, 115V/60Hz, R1270 Refrigerant. -50…200C. 1kW heater. Cooling power 1.26kW at 20C, I/O: USB-B, RS232/RS285, Ethernet; External Pt100, programmable. USB-A data-logging.
SDP
Catalog No. 5000693603
Encompass_Preferred
Refrigerated/heating circulator, MAGIO MS-1200FW, water-cooled, 115V/60Hz, R290 Refrigerant. -50…200C. 1kW heater. Cooling power 1.26kW at 20C 1.11kW at 0C 640W at -20C 390W at -30C 220W at -40C. Adjustable pressure/suction centrifugal pump 16-31LPM, 3.5-13.3 PSI. USB-A data-logging; USB-B, RS232/RS485, Ethernet (Modbus TCP/IP) external communication. Lemosa Pt100 port for external temperature control and monitoring. Color touchscreen UI. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 5...7.5L bath tank. Bath tank opening: 7.1x5.1/5.9 in (WxL/D); Dimensions: 13 x 18.5 x 27.6 in. 97lbs; G3/4" threaded fittings for water cooling.
Catalog No. 50-006-93603 Supplier Julabo USA Inc Supplier No. 9032728.N1.22
Refrigerated/heating circulator, MAGIO MS-1200FW, water-cooled, 115V/60Hz, R290 Refrigerant. -50…200C. 1kW heater. Cooling power 1.26kW at 20C 1.11kW at 0C 640W at -20C 390W at -30C 220W at -40C. Adjustable pressure/suction centrifugal pump 16-31LPM, 3.5-13.3 PSI. USB-A data-logging; USB-B, RS232/RS485, Ethernet (Modbus TCP/IP) external communication. Lemosa Pt100 port for external temperature control and monitoring. Color touchscreen UI. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 5...7.5L bath tank. Bath tank opening: 7.1x5.1/5.9 in (WxL/D); Dimensions: 13 x 18.5 x 27.6 in. 97lbs; G3/4" threaded fittings for water cooling.