Learn More
Julabo USA Inc Refrigerated/heating circulator, JULABO, MAGIO MS-1200FW, water-cooled, 230V/60Hz, R1270 Refrigerant. -50…200C. 2kW heater. Cooling power 1.26kW at 20C, I/O: USB-B, RS232/RS285, Ethernet; External Pt100, programmable. USB-A data-logging.
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'