Learn More
Julabo USA Inc Refrigerated/heating circulator, JULABO, MAGIO MS-449F, 115V/60Hz, R290 Refrigerant. -30…200C. 1kW heater. Cooling power 460W at 20C, I/O: USB-B, RS232/RS285, Ethernet; External Pt100, programmable. USB-A data-logging.
SDP
Catalog No. 5000693614
Encompass_Preferred
Refrigerated/heating circulator, MAGIO MS-449F, 115V/60Hz, R290 Refrigerant. -30…200C. 1kW heater. Cooling power 460W at 20C 37W at 0C 290W at -10C 220W at -20C 80W at -30C. Adjustable pressure/suction centrifugal pump 16-31LPM, 3.5-13.3 PSI. USB-A data-logging; USB-B, RS232/RS485, Ethernet (Modbus TCP/IP) external communication. Lemosa Pt100 port for external temperature control and monitoring. Color touchscreen UI. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 18...26L bath tank. Bath tank opening: 11x13.875.9 in (WxL/D), recommended for internal bath applications; Dimensions: 14.6x 23.2 x 27.2 in. 92.6lbs
Catalog No. 50-006-93614 Supplier Julabo USA Inc Supplier No. 9032716.N1.22
Refrigerated/heating circulator, MAGIO MS-449F, 115V/60Hz, R290 Refrigerant. -30…200C. 1kW heater. Cooling power 460W at 20C 37W at 0C 290W at -10C 220W at -20C 80W at -30C. Adjustable pressure/suction centrifugal pump 16-31LPM, 3.5-13.3 PSI. USB-A data-logging; USB-B, RS232/RS485, Ethernet (Modbus TCP/IP) external communication. Lemosa Pt100 port for external temperature control and monitoring. Color touchscreen UI. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 18...26L bath tank. Bath tank opening: 11x13.875.9 in (WxL/D), recommended for internal bath applications; Dimensions: 14.6x 23.2 x 27.2 in. 92.6lbs