Learn More
Julabo USA Inc DD-200F refrigerated/heating circulator, -20…+200°C, 200W cooling, power, 2kW heating power, internal/external applications, 4L bath tank, adjustable pump 8-23L/min, 1.8-8.7 psi, 8/12 mm hose barbs, USB, Pt100 port, TFT display, 230V/50-60Hz, EA.
SDP
Catalog No. 501539925
Encompass_Preferred
JULABO DYNEO™ DD-200F Refrigerated/heating circulator -20…+200 °C ±0.01 °C; 200W cooling @ 200 °C; 200W cooling @ 20 °C; 170W @ 10 °C; 150W @ 0 °C; 100W @ -10 °C; 20W @ -20 °C; 2 kW heater; 3-4 L bath tank; Bath opening (WxL/D): 5.1x5.9/5.9 inches; adjustable flow pump 8-23L/min; 1.5-8.7 psi with 8 and 12 mm hose barbs; for internal/external applications; Color TFT display with central rotary knob for input. Integrated programmer. Multi-lingual. USB port for PC communication; Pt100 port for external temperature control; Dimensions (WxLxH): 9.1x15.4x25.6 inches; 56.7 pounds. 230V/50-60Hz.
Catalog No. 50-153-9925 Supplier Julabo USA Inc Supplier No. 9021701.33
Add to Cart
Add to Cart
Request a Quote
JULABO DYNEO™ DD-200F Refrigerated/heating circulator -20…+200 °C ±0.01 °C; 200W cooling @ 200 °C; 200W cooling @ 20 °C; 170W @ 10 °C; 150W @ 0 °C; 100W @ -10 °C; 20W @ -20 °C; 2 kW heater; 3-4 L bath tank; Bath opening (WxL/D): 5.1x5.9/5.9 inches; adjustable flow pump 8-23L/min; 1.5-8.7 psi with 8 and 12 mm hose barbs; for internal/external applications; Color TFT display with central rotary knob for input. Integrated programmer. Multi-lingual. USB port for PC communication; Pt100 port for external temperature control; Dimensions (WxLxH): 9.1x15.4x25.6 inches; 56.7 pounds. 230V/50-60Hz.