Learn More
Julabo USA Inc DD-1000F refrigerated/heating circulator, -50…+200°C, 1000W cooling, power, 1kW heating power, internal/external applications, 7.5L bath tank, pump 8-23L/min, 1.8-8.7 psi, 8/12 mm hose barbs, USB; Analog module, Pt100 port, TFT display, 115V/60Hz, EA
SDP
Catalog No. 501539964
Encompass_Preferred
JULABO DYNEO™ DD-1000F Refrigerated/heating circulator -50…+200 °C ±0.01 °C;1000W cooling @ 200 °C; 1000W cooling @ 20 °C; 1000W @ 10 °C; 900W @ 0 °C; 730W @ -10 °C; 500W @ -20 °C; 300W @ -30°C; 130W @ -40°C; 1 kW heater; 5-7.5 L bath tank; Bath opening (WxL/D): 8.7x5.9/5.9 inches; adjustable flow pump 8-23L/min; 1.5-8.7 psi with 8 and 12 mm hose barbs; for internal/external applications; Analog module for external voltage/current control. Color TFT display with central rotary knob for input. Integrated programmer. Multi-lingual. USB port for PC integration + Analog module; Pt100 port for external temperature control; Dimensions (WxLxH): 16.5x19.3x27.6 inches; 112.9 pounds. 115V/60Hz.
Catalog No. 50-153-9964 Supplier Julabo USA Inc Supplier No. 9021707.02.A
Add to Cart
Add to Cart
Request a Quote
JULABO DYNEO™ DD-1000F Refrigerated/heating circulator -50…+200 °C ±0.01 °C;1000W cooling @ 200 °C; 1000W cooling @ 20 °C; 1000W @ 10 °C; 900W @ 0 °C; 730W @ -10 °C; 500W @ -20 °C; 300W @ -30°C; 130W @ -40°C; 1 kW heater; 5-7.5 L bath tank; Bath opening (WxL/D): 8.7x5.9/5.9 inches; adjustable flow pump 8-23L/min; 1.5-8.7 psi with 8 and 12 mm hose barbs; for internal/external applications; Analog module for external voltage/current control. Color TFT display with central rotary knob for input. Integrated programmer. Multi-lingual. USB port for PC integration + Analog module; Pt100 port for external temperature control; Dimensions (WxLxH): 16.5x19.3x27.6 inches; 112.9 pounds. 115V/60Hz.