Learn More
Electron Microscopy Sciences EMS Oscillating Tissue Slicer Refrigeration Retrofit
SDP
Catalog No. 5025771
Encompass_Preferred
Provide refrigeration capabilities to the 5000 Tissue Slicer. The controller three digit LED displays the current temperature of the bath medium in degrees C. The temperature is maintained within 0.5°C. The temperature range is between 0-10°C.
Catalog No. 50-257-71 Supplier Electron Microscopy Sciences Supplier No. 93226
Provide refrigeration capabilities to the 5000 Tissue Slicer. The controller three digit LED displays the current temperature of the bath medium in degrees C. The temperature is maintained within 0.5°C. The temperature range is between 0-10°C.