Learn More
VWR International REFRIGERATOR/FRZER THERMOMETER
Catalog No. NC1920350
Encompass_Preferred
Refrigerator/Freezer Thermometer–50 to 70°C (–58 to 158°F)Bottle Probe
Catalog No. NC1920350 Supplier VWR International Supplier No. 61161364
Refrigerator/Freezer Thermometer–50 to 70°C (–58 to 158°F)Bottle Probe