Learn More
Thomas Scientific 30 cu. ft. Standard Explosion Proof Refrigerator
SDP
Catalog No. 501457312
Encompass_Preferred
Units come standard with mechanical thermostat and leveling legs. Exterior Color: White Refrigerant: R134a (except 1182V49 & 1182V50 use R404a) Defrost: Cycle Temperature Range: 1 to 10°C (34 to 50°F) UL/C-UL listed 1 Years Parts & Labor
This item is not returnable. View return policy
Catalog No. 50-145-7312 Supplier Thomas Scientific Supplier No. 1182V49
Add to Cart
Add to Cart
Request a Quote
Units come standard with mechanical thermostat and leveling legs. Exterior Color: White Refrigerant: R134a (except 1182V49 & 1182V50 use R404a) Defrost: Cycle Temperature Range: 1 to 10°C (34 to 50°F) UL/C-UL listed 1 Years Parts & Labor