Learn More
Darwin Chambers Company REFRIGERATOR STABILITY 47 CUFT
SDP
Supplier:  Darwin Chambers Company ST055AA
Encompass_Preferred
The ST055 is a 50 cubic feet double door stability refrigerator. Temperature range is 2°C to 15°C. This unit comes with 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. This model features solid state controlled, hot gas bypass refrigeration control method with electric heat assisted refrigeration. The ST series chambers control well within the specifications required by the FDA and ICH for GMP studies.
Catalog No. 50-1251789
Only null left
Add to Cart
Request a Quote