Learn More
Darwin Chambers Company Single Door Stability Refrigerator 9 cu. ft.
SDP
Catalog No. 501893925
Encompass_Preferred
The ST011 is a 11.6 cubic feet benchtop stability refrigerator. Temperature range is 2°C to 15°C. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. This model features solid state controlled, hot gas bypass refrigeration control method with electric heat assisted refrigeration. The ST series chambers control well within the specifications required by the FDA and ICH for GMP studies.
Catalog No. 50-189-3925 Supplier Darwin Chambers Company Supplier No. ST011
Add to Cart
Add to Cart
Request a Quote
The ST011 is a 11.6 cubic feet benchtop stability refrigerator. Temperature range is 2°C to 15°C. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. This model features solid state controlled, hot gas bypass refrigeration control method with electric heat assisted refrigeration. The ST series chambers control well within the specifications required by the FDA and ICH for GMP studies.