Learn More
Darwin Chambers Company Triple Door Stability Refrigerator 66 cu. ft.
SDP
Catalog No. 501893929
Encompass_Preferred
The ST084-SS is a 76.2 cubic feet triple door stability refrigerator. Temperature range is 2°C to 15°C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. This model features solid state controlled, hot gas bypass refrigeration control method with electric heat assisted refrigeration. The ST series chambers control well within the specifications required by the FDA and ICH for GMP studies.
Catalog No. 50-189-3929 Supplier Darwin Chambers Company Supplier No. ST084SS
Add to Cart
Add to Cart
Request a Quote
The ST084-SS is a 76.2 cubic feet triple door stability refrigerator. Temperature range is 2°C to 15°C. This unit comes with 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. This model features solid state controlled, hot gas bypass refrigeration control method with electric heat assisted refrigeration. The ST series chambers control well within the specifications required by the FDA and ICH for GMP studies.