Learn More
Euthanex Refresh Charcoal Canister 8 set
Catalog No. NC0123825
Encompass_Preferred
Less waste, increased convenience and significant cost savings. Third party testing compared the E-Z Anesthesia ReFresh Filter with other commonly used filters. The facility introduced 2% isoflurane in medical air into each filter at a 1 liter/min flow rate. The test was terminated at the manufacturers recommended saturation point for each filter. The ReFresh filter lasted over two times as long as the other filters with zero waste gas emissions. Sold in cases of eight (8).
Catalog No. NC0123825 Supplier Euthanex Supplier No. EZ258
Less waste, increased convenience and significant cost savings. Third party testing compared the E-Z Anesthesia ReFresh Filter with other commonly used filters. The facility introduced 2% isoflurane in medical air into each filter at a 1 liter/min flow rate. The test was terminated at the manufacturers recommended saturation point for each filter. The ReFresh filter lasted over two times as long as the other filters with zero waste gas emissions. Sold in cases of eight (8).