Learn More
Medchemexpress LLC REG-3 gamma/REG3G H 100ug
SDP
Catalog No. 5000197205
Encompass_Preferred
Recombinant human REG-3 gamma (REG3G) protein produced in HEK293 cells and supplied as a lyophilized powder with a C-terminal 6xHis tag. Designed for biochemical and functional assays, bactericidal activity studies, and antibody or assay development, the product offers >95% purity, low endotoxin, and a formulation that is ready to reconstitute for downstream applications.
  • Expressed in HEK293 cells with a C-terminal His tag.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4 for stability and handling.
  • Stable when stored at -20°C for up to two years; aliquot and freeze at -80°C for extended storage.
  • Available in multiple sizes, including 100 μg, to suit experimental needs.
Catalog No. 50-001-97205 Supplier Medchemexpress LLC Supplier No. HYP71256100UG
Recombinant human REG-3 gamma (REG3G) protein produced in HEK293 cells and supplied as a lyophilized powder with a C-terminal 6xHis tag. Designed for biochemical and functional assays, bactericidal activity studies, and antibody or assay development, the product offers >95% purity, low endotoxin, and a formulation that is ready to reconstitute for downstream applications.
  • Expressed in HEK293 cells with a C-terminal His tag.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4 for stability and handling.
  • Stable when stored at -20°C for up to two years; aliquot and freeze at -80°C for extended storage.
  • Available in multiple sizes, including 100 μg, to suit experimental needs.