Learn More
Research Products International Corp Reagent Reservoir RL12, S, 10/case
SDP
Catalog No. 502229660
Encompass
Ideal Reagent Reservoir RL12 allows user to perform serial dilutions and pipetting of different reagents with 8 or 12 multi-channel pipettor. Channels are fabricated to allow complete removal of sample from each well. Individual channels are clearly marked and hold 5 ml of solution. Rigid design offers stability, ensures secure stackability and includes fitted lid that minimizes evaporation. Made in the USA.
12 Channel Reservoir with lid
Autoclavable polyporpylene
Catalog No. 50-222-9660 Supplier Research Products International Corp Supplier No. 212350S
Ideal Reagent Reservoir RL12 allows user to perform serial dilutions and pipetting of different reagents with 8 or 12 multi-channel pipettor. Channels are fabricated to allow complete removal of sample from each well. Individual channels are clearly marked and hold 5 ml of solution. Rigid design offers stability, ensures secure stackability and includes fitted lid that minimizes evaporation. Made in the USA.
12 Channel Reservoir with lid
Autoclavable polyporpylene