Learn More
ERA WATERS CORP REG VOLATILES QC POTABLE WATR
SDP
Catalog No. NC0291926
Encompass
One 2 mL flame-sealed ampule yields in excess of 200 mL after dilution. Contains all of the analytes below at 2-50 ug/L.
Catalog No. NC0291926 Supplier ERA WATERS CORP Supplier No. 703
One 2 mL flame-sealed ampule yields in excess of 200 mL after dilution. Contains all of the analytes below at 2-50 ug/L.